Dysprosium powder, -40 mesh, 99.9% (REO)
Dysprosium powder, -40 mesh, 99.9% (REO)
Dysprosium powder, -40 mesh, 99.9% (REO)
Thermo Scientific Chemicals

Dysprosium powder, -40 mesh, 99.9% (REO)

CAS: 7429-91-6 | Dy | 162.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 012897.14
also known as 012897-14
Price (USD)
520.65
Online Exclusive
578.00
Save 57.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
520.65
Online Exclusive
578.00
Save 57.35 (10%)
Each
Chemical Identifiers
CAS7429-91-6
IUPAC Namedysprosium
Molecular FormulaDy
InChI KeyKBQHZAAAGSGFKK-UHFFFAOYSA-N
SMILES[Dy]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only