Ytterbium(III) nitrate hydrate, 99.9% (REO)
Thermo Scientific Chemicals

Ytterbium(III) nitrate hydrate, 99.9% (REO)

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 012901.09
also known as 012901-09
Price (USD)
58.30
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
58.30
Each
Specifications
Assay Percent Notes(REO)
Chemical Name or MaterialYtterbium(III) nitrate hydrate
ColorWhite
DOT InformationTransport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECS Number237-384-6
View more
Ytterbium(III) nitrate hydrate is used as a precursor of nanoscale coatings of carbon composites. The ytterbium +3 ion is used as a doping material in solid state lasers and double clad fiber lasers. It is also used to make yttrium-based surfactant mesophases, which has adsorbing as well as optically functional properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only