Silver two part conductive adhesive, EXP. XX/XX Resin and Hardener
Thermo Scientific Chemicals

Silver two part conductive adhesive, EXP. XX/XX Resin and Hardener

A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number 012942.03
also known as 012942-03
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
93.80
Save 14.15 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
93.80
Save 14.15 (15%)
Each
Specifications
Chemical Name or MaterialSilver two part conductive adhesive
MDL NumberMFCD00003397
Name NoteEXP. x X/XX Resin and Hardener
Recommended StorageAmbient temperatures
Storage Note 1
  • Limited shelf life, 60 days after shipment
View more
A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Limited shelf life, 60 days after shipment
RUO – Research Use Only