Bright Brushing Gold
Thermo Scientific Chemicals

Bright Brushing Gold

 
Have Questions?
Catalog NumberQuantity
012943.04
also known as 012943-04
2 g
Catalog number 012943.04
also known as 012943-04
Price (USD)
-
Quantity:
2 g
Request bulk or custom format
Specifications
Boiling Point156°C
Chemical Name or MaterialBright Brushing Gold
ColorRed to Brown
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point33°C (91°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only