Iron lumps, vacuum remelted, low Oxygen, 99.99% (metals basis)
Iron lumps, vacuum remelted, low Oxygen, 99.99% (metals basis)
Iron lumps, vacuum remelted, low Oxygen, 99.99% (metals basis)
Thermo Scientific Chemicals

Iron lumps, vacuum remelted, low Oxygen, 99.99% (metals basis)

CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 012958.22
also known as 012958-22
Price (USD)
688.65
Online Exclusive
765.00
Save 76.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
688.65
Online Exclusive
765.00
Save 76.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7439-89-6
ChEBICHEBI:82664
Chemical Name or MaterialIron lumps, vacuum remelted
EINECS Number231-096-4
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only