Thermo Scientific Chemicals

Sodium fluoride, Puratronic™, 99.995% (metals basis excluding K), K =100ppm, Thermo Scientific Chemicals

Catalog number: 012964.18
50 g, Each
Thermo Scientific Chemicals

Sodium fluoride, Puratronic™, 99.995% (metals basis excluding K), K =100ppm, Thermo Scientific Chemicals

Catalog number: 012964.18
50 g, Each
Quantity

Chemical Identifiers

CAS
7681-49-4
IUPAC Name
sodium fluoride
Molecular Formula
FNa
InChI Key
PUZPDOWCWNUUKD-UHFFFAOYSA-M
SMILES
[F-].[Na+]
Total Metal Impurities excluding K
0.005% max.
Impurity content
K ≈60ppm

Description

It is used as a steel degreasing agent and a flux. It is used in heat treating salt compositions, tin-electroplating, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It has significant applications in organic synthesis including its halide-exchange with acyl chloride, sulfur chlorides, and phosphorus chloride, desilylation of silyl-protected OH groups, synthesis of fluorocarbons (Finkelstein reaction), and also in metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text