Ammonium bromide, Puratronic™, 99.999% (metals basis)
Ammonium bromide, Puratronic™, 99.999% (metals basis)
Ammonium bromide, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Ammonium bromide, Puratronic™, 99.999% (metals basis)

CAS: 12124-97-9 | BrH4N | 97.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 012981.06
also known as 012981-06
Price (USD)
43.65
Special offer
Online exclusive
Ends: 28-Dec-2026
50.60
Save 6.95
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 28-Dec-2026
50.60
Save 6.95
Each
Chemical Identifiers
CAS12124-97-9
IUPAC Nameammonium bromide
Molecular FormulaBrH4N
InChI KeySWLVFNYSXGMGBS-UHFFFAOYSA-N
SMILES[NH4+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Ammonium bromide is used in pharmaceuticals for its bromide ion. It is used for a variety of analytical chemistry applications. It is also used for photography in films, plates and papers; in fireproofing of wood; in lithography and process engraving and as corrosion inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only