Lithium sulfate hydrate, Puratronic™, 99.996% (metals basis)
Lithium sulfate hydrate, Puratronic™, 99.996% (metals basis)
Lithium sulfate hydrate, Puratronic™, 99.996% (metals basis)
Thermo Scientific Chemicals

Lithium sulfate hydrate, Puratronic™, 99.996% (metals basis)

CAS: 10377-48-7 | Li2O4S | 109.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 012983.06
also known as 012983-06
Price (USD)
44.50
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
44.50
Each
Chemical Identifiers
CAS10377-48-7
IUPAC Namedilithium(1+) sulfate
Molecular FormulaLi2O4S
InChI KeyINHCSSUBVCNVSK-UHFFFAOYSA-L
SMILES[Li+].[Li+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.004% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only