Strontium acetate hydrate, Puratronic™, 99.9965% (metals basis)
Strontium acetate hydrate, Puratronic™, 99.9965% (metals basis)
Strontium acetate hydrate, Puratronic™, 99.9965% (metals basis)
Thermo Scientific Chemicals

Strontium acetate hydrate, Puratronic™, 99.9965% (metals basis)

CAS: 543-94-2 | C4H6O4Sr | 205.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 012990.06
also known as 012990-06
Price (USD)
81.65
Special offer
Online exclusive
Ends: 28-Dec-2026
96.30
Save 14.65
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
81.65
Special offer
Online exclusive
Ends: 28-Dec-2026
96.30
Save 14.65
Each
Chemical Identifiers
CAS543-94-2
IUPAC Namestrontium(2+) diacetate
Molecular FormulaC4H6O4Sr
InChI KeyRXSHXLOMRZJCLB-UHFFFAOYSA-L
SMILES[Sr++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0035% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only