Mercury(II) sulfide, black, Puratronic™, 99.999% (metals basis)
Mercury(II) sulfide, black, Puratronic™, 99.999% (metals basis)
Mercury(II) sulfide, black, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Mercury(II) sulfide, black, Puratronic™, 99.999% (metals basis)

CAS: 1344-48-5 | HgS | 232.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012992.06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1344-48-5
IUPAC Namemercury(2+) sulfanediide
Molecular FormulaHgS
InChI KeyLGZXYFMMLRYXLK-UHFFFAOYSA-N
SMILES[S--].[Hg++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Mercury(II) sulfide is used in paint, sealing wax and preservatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only