Calcium oxalate monohydrate, Puratronic™, 99.9985% (metals basis), (excluding 25ppm max alkali earths)
Calcium oxalate monohydrate, Puratronic™, 99.9985% (metals basis), (excluding 25ppm max alkali earths)
Calcium oxalate monohydrate, Puratronic™, 99.9985% (metals basis), (excluding 25ppm max alkali earths)
Thermo Scientific Chemicals

Calcium oxalate monohydrate, Puratronic™, 99.9985% (metals basis), (excluding 25ppm max alkali earths)

CAS: 5794-28-5 | C2H2CaO5 | 146.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 013007.14
also known as 013007-14
Price (USD)
222.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
222.00
Each
Chemical Identifiers
CAS5794-28-5
IUPAC Namecalcium hydrate oxalate
Molecular FormulaC2H2CaO5
InChI KeyLQHWSGSWNOHVHO-UHFFFAOYSA-L
SMILESO.[Ca++].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Comment25ppm max alkali earths
Total Metal Impurities≤0.0015%
FormGranules/powder
Appearance (Color)White
Calcium oxalate monohydrate is used as a precursor in the production of oxalic acid, organic oxalates, ceramic glazes, and for rare-earth-metal separation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only