Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Thermo Scientific™
Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Perform virtually any electrochemistry analysis with these conductivity standards, solutions and accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
013017Replacement Flow Cell
1010001Calibration Resistor Kit
Catalog number 013017
Price (MXN)
-
Type:
Replacement Flow Cell
Thermo Scientific™Orion&trade Glass Flow Cell is designed for accurate measurement of low-conductivity solutions. When attached directly to the probe, it minimizes the risk of air introduction into the sample during analysis, helping to ensure reliable results.
The Glass Flow Cell is compatible with the Thermo Scientific&tradeOrion&trade 2-Cell Ultrapure Conductivity Probe (Cat. No. 013016MD).
The Thermo Scientific&trade Orion&tradeConductivity Resistor Calibration Kit is designed to verify the performance of Orion&tradeconductivity meters below
Compatible with the following Thermo Scientific™ Orion™ conductivity meters:
  • Versa Star Pro Conductivity Bench Meters (VSTAR20, VSTAR50, VSTAR90, VSTAR91, VSTAR-CND)
  • Pro Star Conductivity Bench Meters (PSTAR2120, PSTAR2150, PSTAR2180)
  • Lab Star Conductivity Bench Meters (LSTAR1120)
  • Legacy Orion Star A210 Conductivity Bench Meters (STARA2120, STARA2150)
  • Legacy Orion Star A110 Conductivity Bench Meters (STARA1120)
Specifications
DescriptionReplacement Glass Flow Cell
NIST TraceableYes
For Use With (Equipment)Conductivity Probes (013016MD, 013016A and 013016D)
TypeReplacement Flow Cell
Unit SizeEach