Arsenic polycrystalline lump, 2-8mm (0.08-0.3in), Puratronic™, 99.99999+% (metals basis)
Arsenic polycrystalline lump, 2-8mm (0.08-0.3in), Puratronic™, 99.99999+% (metals basis)
Arsenic polycrystalline lump, 2-8mm (0.08-0.3in), Puratronic™, 99.99999+% (metals basis)
Arsenic polycrystalline lump, 2-8mm (0.08-0.3in), Puratronic™, 99.99999+% (metals basis)
Thermo Scientific Chemicals

Arsenic polycrystalline lump, 2-8mm (0.08-0.3in), Puratronic™, 99.99999+% (metals basis)

CAS: 7440-38-2 | As | 74.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
25 g
100 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 013044.A1
also known as 013044-A1
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS7440-38-2
IUPAC Namearsenic
Molecular FormulaAs
InChI KeyRQNWIZPPADIBDY-UHFFFAOYSA-N
SMILES[As]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metalsless than 0.00001% trace metal impurities
Appearance (Color)Grey
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only