Calcium tungsten oxide, 98%
Calcium tungsten oxide, 98%
Calcium tungsten oxide, 98%
Thermo Scientific Chemicals

Calcium tungsten oxide, 98%

CAS: 7790-75-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 013051.36
also known as 013051-36
Price (USD)
306.65
Online Exclusive
341.00
Save 34.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
306.65
Online Exclusive
341.00
Save 34.35 (10%)
Each
Chemical Identifiers
CAS7790-75-2
Molecular FormulaCaO4W
InChI KeyFDMFQOCGNBYKPY-UHFFFAOYSA-N
SMILES[Ca++].[O-][W]([O-])(=O)=O
Molecular Weight (g/mol)287.91
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pink to pale brown
FormPowder
Assay (unspecified)≥97.5%
Calcium tungsten oxide is used as a luminophore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only