Dodecacarbonyltriiron, 96% (dry wt.), stab. with 5-10% methanol
Dodecacarbonyltriiron, 96% (dry wt.), stab. with 5-10% methanol
Dodecacarbonyltriiron, 96% (dry wt.), stab. with 5-10% methanol
Thermo Scientific Chemicals

Dodecacarbonyltriiron, 96% (dry wt.), stab. with 5-10% methanol

Reducing agent (reduces NO2), catalyst for alkyne trimerization | CAS: 17685-52-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 013059.04
also known as 013059-04
Price (USD)
35.70
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
35.70
Each
Chemical Identifiers
CAS17685-52-8
IUPAC Namedodecakis(methanidylidyneoxidanium) triiron
Molecular FormulaC12Fe3O12
InChI KeyHLYRMDDXFDINCB-UHFFFAOYSA-N
SMILES[Fe].[Fe].[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormCrystals or powder
Assay from Supplier's CofA≥95%
It eliminates sulfur from substituted thiophene complexes, which leads to ferroles. Access to new areas of organometallic chemistry is promising using the reaction of Fe3(CO)12 with tellurium-nitrogen heterocycles such as benzoisotellurazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only