Octacarbonyldicobalt, stab. with 1-5% hexane
Octacarbonyldicobalt, stab. with 1-5% hexane
Octacarbonyldicobalt, stab. with 1-5% hexane
Thermo Scientific Chemicals

Octacarbonyldicobalt, stab. with 1-5% hexane

As a catalyst in oxo process, and in high-purity cobalt salts | CAS: 10210-68-1
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
25 g
100 g
Catalog number 013060.14
also known as 013060-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS10210-68-1
Molecular FormulaC8Co2O8
InChI KeyZUDHYLQHRXCZGU-UHFFFAOYSA-N
SMILES[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].O=[C-]1[Co][C-](=O)[Co++]1
Molecular Weight (g/mol)341.95
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps
Appearance (Color)Red to orange to brown
Assay (unspecified)33 to 36 % Co
As a catalyst in oxo process, and in high-purity cobalt saltsOctacarbonyldicobalt is used as a catalyst for hydroformylation. It plays an important role in promoting both the Nicholas reaction and the Pauson-Khand reaction. It acts as a reagent and catalyst in organometallic chemistry and organic synthesis. It is involved as an alkyne protecting group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only