Nonacarbonyldiiron, 99%
Nonacarbonyldiiron, 99%
Nonacarbonyldiiron, 99%
Thermo Scientific Chemicals

Nonacarbonyldiiron, 99%

CAS: 15321-51-4 | C9H3Fe2O9 | 366.804 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 013062.18
also known as 013062-18
Price (USD)
412.65
Online Exclusive
458.00
Save 45.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
412.65
Online Exclusive
458.00
Save 45.35 (10%)
Each
Chemical Identifiers
CAS15321-51-4
IUPAC Nameiron(3+) hexakis(methanidylidyneoxidanium) tris(oxomethanide) iron
Molecular FormulaC9H3Fe2O9
InChI KeyCGJSNKHFIBINMR-UHFFFAOYSA-N
SMILES[Fe].[Fe+3].[CH-]=O.[CH-]=O.[CH-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Appearance (Color)Yellow to orange
Assay from Suppliers CofA≥98.5%
It is an important reagent in organometallic chemistry and of occasional use in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only