Tin(IV) fluoride, 99% (metals basis)
Tin(IV) fluoride, 99% (metals basis)
Tin(IV) fluoride, 99% (metals basis)
Thermo Scientific Chemicals

Tin(IV) fluoride, 99% (metals basis)

Friedel-Crafts catalyst | CAS: 7783-62-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 013063.04
also known as 013063-04
Price (USD)
85.30
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
85.30
Each
Chemical Identifiers
CAS7783-62-2
IUPAC Nametin(4+) tetrafluoride
Molecular FormulaF4Sn
InChI KeyYUOWTJMRMWQJDA-UHFFFAOYSA-J
SMILES[F-].[F-].[F-].[F-].[Sn+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
It is used as Friedel-Crafts catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only