Sodium hexafluoroantimonate(V), 99.5%
Sodium hexafluoroantimonate(V), 99.5%
Sodium hexafluoroantimonate(V), 99.5%
Thermo Scientific Chemicals

Sodium hexafluoroantimonate(V), 99.5%

CAS: 16925-25-0 | F6NaSb | 258.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 013066.22
also known as 013066-22
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS16925-25-0
Molecular FormulaF6NaSb
InChI KeyZJLVIRKMUXZEDU-UHFFFAOYSA-N
SMILES[F].[F].[F].[F].[F].[F].[Na+].[Sb]
Molecular Weight (g/mol)258.74
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥43.8% Fluoride
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only