Copper(II) fluoride dihydrate
Copper(II) fluoride dihydrate
Copper(II) fluoride dihydrate
Thermo Scientific Chemicals

Copper(II) fluoride dihydrate

Flux in casting gray iron | CAS: 13454-88-1 | CuF2H4O2 | 137.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 013076.18
also known as 013076-18
Price (USD)
51.65
Online Exclusive
57.40
Save 5.75 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
51.65
Online Exclusive
57.40
Save 5.75 (10%)
Each
Chemical Identifiers
CAS13454-88-1
IUPAC Namecopper(2+) dihydrate difluoride
Molecular FormulaCuF2H4O2
InChI KeyCTRICEWDSISGPV-UHFFFAOYSA-L
SMILESO.O.[F-].[F-].[Cu++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)27.3% F min
Copper(II) fluoride can also be used to form fluorobenzene from benzene, hydrochloric acid and oxygen. Using a metal fluoride that can successfully oxidize with a methyl bond. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only