Titanium(III) fluoride
Titanium(III) fluoride
Titanium(III) fluoride
Thermo Scientific Chemicals

Titanium(III) fluoride

CAS: 13470-08-1
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 013079.18
also known as 013079-18
Price (USD)
311.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
311.00
Each
Chemical Identifiers
CAS13470-08-1
IUPAC Nametitanium(3+) trifluoride
Molecular FormulaF3Ti
InChI KeyNLPMQGKZYAYAFE-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Ti+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)53.26% F min

It is used as a laboratory chemicals & for manufacture of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only