Tungsten(IV) selenide, 99.8% (metals basis)
Tungsten(IV) selenide, 99.8% (metals basis)
Tungsten(IV) selenide, 99.8% (metals basis)
Thermo Scientific Chemicals

Tungsten(IV) selenide, 99.8% (metals basis)

CAS: 12067-46-8 | Se2W | 341.782 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 013084.18
also known as 013084-18
Price (USD)
324.65
Online Exclusive
361.00
Save 36.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
324.65
Online Exclusive
361.00
Save 36.35 (10%)
Each
Chemical Identifiers
CAS12067-46-8
IUPAC Namediselanylidenetungsten
Molecular FormulaSe2W
InChI KeyROUIDRHELGULJS-UHFFFAOYSA-N
SMILES[Se]=[W]=[Se]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size10-20 Microns average
Total Metal Impurities≤0.2% (2000 ppm)
FormPowder
Appearance (Color)Dark gray
Tungsten(IV) selenide is used as an important compound in electrochemical solar cells. It acts as a stable semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only