Titanium silicide, 99.5% (metals basis)
Titanium silicide, 99.5% (metals basis)
Titanium silicide, 99.5% (metals basis)
Thermo Scientific Chemicals

Titanium silicide, 99.5% (metals basis)

CAS: 12039-83-7 | Si2Ti | 104.037 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 013088.30
also known as 013088-30
Price (USD)
423.65
Special offer
Online exclusive
Ends: 30-Jun-2026
498.00
Save 74.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
423.65
Special offer
Online exclusive
Ends: 30-Jun-2026
498.00
Save 74.35 (15%)
Each
Chemical Identifiers
CAS12039-83-7
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Titanium silicide is used in the semiconductor industry. It is also used in the fabrication of transistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only