Tantalum nitride, 99.5% (metals basis)
Tantalum nitride, 99.5% (metals basis)
Tantalum nitride, 99.5% (metals basis)
Thermo Scientific Chemicals

Tantalum nitride, 99.5% (metals basis)

CAS: 12033-62-4 | NTa | 194.955 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 013093.18
also known as 013093-18
Price (USD)
294.65
Special offer
Online exclusive
Ends: 30-Jun-2026
347.00
Save 52.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
294.65
Special offer
Online exclusive
Ends: 30-Jun-2026
347.00
Save 52.35 (15%)
Each
Chemical Identifiers
CAS12033-62-4
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Tantalum nitride is used to create barrier or glue layers between copper, or other conductive metals, and dielectric insulator films such as thermal oxides. These films are deposited on top of silicon wafers during the manufacture of integrated circuits, to create thin film surface mount resistors and has other electronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only