Niobium silicide, 99.85% (metals basis)
Niobium silicide, 99.85% (metals basis)
Niobium silicide, 99.85% (metals basis)
Thermo Scientific Chemicals

Niobium silicide, 99.85% (metals basis)

CAS: 12034-80-9 | NbSi2 | 149.076 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 013100.22
also known as 013100-22
Price (USD)
419.65
Online Exclusive
466.00
Save 46.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
419.65
Online Exclusive
466.00
Save 46.35 (10%)
Each
Chemical Identifiers
CAS12034-80-9
IUPAC Name(silanylideneniobio)silane
Molecular FormulaH4NbSi2
InChI KeyOVICFFSAIBBAPC-UHFFFAOYSA-N
SMILES[SiH2]=[Nb]=[SiH2]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.15% max.
Niobium silicide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only