Nickel(II) telluride, 99.9% (metals basis excluding Co), Co 0.1-1%
Nickel(II) telluride, 99.9% (metals basis excluding Co), Co 0.1-1%
Nickel(II) telluride, 99.9% (metals basis excluding Co), Co 0.1-1%
Thermo Scientific Chemicals

Nickel(II) telluride, 99.9% (metals basis excluding Co), Co 0.1-1%

CAS: 12142-88-0 | NiTe | 186.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 013105.18
also known as 013105-18
Price (USD)
339.65
Online Exclusive
377.00
Save 37.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
339.65
Online Exclusive
377.00
Save 37.35 (10%)
Each
Chemical Identifiers
CAS12142-88-0
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
CommentCo 0.1-1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only