Cobalt(II) sulfide, 99.5% (metals basis)
Cobalt(II) sulfide, 99.5% (metals basis)
Cobalt(II) sulfide, 99.5% (metals basis)
Cobalt(II) sulfide, 99.5% (metals basis)
Thermo Scientific Chemicals

Cobalt(II) sulfide, 99.5% (metals basis)

CAS: 1317-42-6 | CoS | 90.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 013114.14
also known as 013114-14
Price (USD)
101.65
Online Exclusive
113.00
Save 11.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
101.65
Online Exclusive
113.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS1317-42-6
IUPAC Nameλ²-cobalt(2+) sulfanediide
Molecular FormulaCoS
InChI KeyINPLXZPZQSLHBR-UHFFFAOYSA-N
SMILES[S--].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.

Used in casting, smelting of metal, hard alloys. Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only