Cadmium iodide, 99.5% (metals basis)
Cadmium iodide, 99.5% (metals basis)
Cadmium iodide, 99.5% (metals basis)
Thermo Scientific Chemicals

Cadmium iodide, 99.5% (metals basis)

CAS: 7790-80-9 | CdI2 | 366.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 013125.14
also known as 013125-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7790-80-9
IUPAC Namecadmium(2+) diiodide
Molecular FormulaCdI2
InChI KeyOKIIEJOIXGHUKX-UHFFFAOYSA-L
SMILES[Cd++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
It is used in lithography, photography, electroplating, as an intermediate and in the manufacturing of phosphors, lubricants, electrode position of Cadmium,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In photography, process engraving and lithography, analytical chemistry, electrodeposition of Cadmium, manufacturing of phosphors, lubricants
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only