Bismuth(III) selenide, Vacuum Deposition Grade, 99.995% (metals basis)
Bismuth(III) selenide, Vacuum Deposition Grade, 99.995% (metals basis)
Bismuth(III) selenide, Vacuum Deposition Grade, 99.995% (metals basis)
Thermo Scientific Chemicals

Bismuth(III) selenide, Vacuum Deposition Grade, 99.995% (metals basis)

In semiconductor research, multilayer thin-film, magnetoresistant film | CAS: 12068-69-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 013126.09
also known as 013126-09
Price (USD)
88.65
Online Exclusive
98.20
Save 9.55 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
88.65
Online Exclusive
98.20
Save 9.55 (10%)
Each
Chemical Identifiers
CAS12068-69-8
IUPAC Namedibismuth(3+) triselandiide
Molecular FormulaBi2Se3
InChI KeyFBGGJHZVZAAUKJ-UHFFFAOYSA-N
SMILES[Se--].[Se--].[Se--].[Bi+3].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPieces, 1-6 mm
Total Metal Impurities≤0.005%
In semiconductor research, multilayer thin-film, magnetoresistant film.Bismuth(III) selenide is used as a semiconductor and thermoelectric material. It is also used in semiconductor research. Further, it finds applications magnetoresistant film and semiconductor research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only