Samarium(III) carbonate hydrate, REacton™, 99.99% (REO)
Samarium(III) carbonate hydrate, REacton™, 99.99% (REO)
Samarium(III) carbonate hydrate, REacton™, 99.99% (REO)
Samarium(III) carbonate hydrate, REacton™, 99.99% (REO)
Samarium(III) carbonate hydrate, REacton™, 99.99% (REO)
Samarium(III) carbonate hydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Samarium(III) carbonate hydrate, REacton™, 99.99% (REO)

CAS: 38245-37-3 | C3O9Sm2 | 480.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 013177.14
also known as 013177-14
Price (USD)
67.65
Special offer
Online exclusive
Ends: 28-Dec-2026
79.80
Save 12.15
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 28-Dec-2026
79.80
Save 12.15
Each
Chemical Identifiers
CAS38245-37-3
IUPAC Namedisamarium(3+) tricarbonate
Molecular FormulaC3O9Sm2
InChI KeyQCZFMLDHLOYOQJ-UHFFFAOYSA-H
SMILES[Sm+3].[Sm+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only