Europium(III) sulfate octahydrate, REacton™, 99.99% (REO)
Europium(III) sulfate octahydrate, REacton™, 99.99% (REO)
Europium(III) sulfate octahydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Europium(III) sulfate octahydrate, REacton™, 99.99% (REO)

CAS: 10031-55-7 | Eu2H16O20S3 | 736.216 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 013210.06
also known as 013210-06
Price (USD)
115.65
Special offer
Online exclusive
Ends: 30-Jun-2026
136.00
Save 20.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
115.65
Special offer
Online exclusive
Ends: 30-Jun-2026
136.00
Save 20.35 (15%)
Each
Chemical Identifiers
CAS10031-55-7
IUPAC Namedieuropium(3+) octahydrate trisulfate
Molecular FormulaEu2H16O20S3
InChI KeyPGDXJFDXKJLVTJ-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.O.O.[Eu+3].[Eu+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only