Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO)
Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO)
Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO)

CAS: 14637-88-8 | C15H21DyO6 | 459.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 013219.14
also known as 013219-14
Price (USD)
160.65
Online Exclusive
178.00
Save 17.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
160.65
Online Exclusive
178.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS14637-88-8
IUPAC Namedysprosium(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21DyO6
InChI KeyVABPDCUJKLETFM-UHFFFAOYSA-N
SMILES[Dy+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only