Rhenium(III) chloride
Rhenium(III) chloride
Rhenium(III) chloride
Thermo Scientific Chemicals

Rhenium(III) chloride

CAS: 13569-63-6 | Cl3Re | 292.557 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
Catalog number 013230.03
also known as 013230-03
Price (USD)
481.65
Special offer
Online exclusive
Ends: 30-Jun-2026
567.00
Save 85.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
481.65
Special offer
Online exclusive
Ends: 30-Jun-2026
567.00
Save 85.35 (15%)
Each
Chemical Identifiers
CAS13569-63-6
IUPAC Namerhenium(3+) trichloride
Molecular FormulaCl3Re
InChI KeyLOIHSHVELSAXQN-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Re+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.5%
FormPowder
It is used as a starting material for synthesis of rhenium complexes. ReCl3 can react with peralkylated benzene-metal complexes that act as electron reservoirs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only