Gallium(III) sulfide, 99.99% (metals basis)
Gallium(III) sulfide, 99.99% (metals basis)
Gallium(III) sulfide, 99.99% (metals basis)
Thermo Scientific Chemicals

Gallium(III) sulfide, 99.99% (metals basis)

CAS: 12024-22-5 | Ga2S3 | 235.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
10 g
Catalog number 013387.09
Price (USD)
384.65
Online Exclusive
427.00
Save 42.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
384.65
Online Exclusive
427.00
Save 42.35 (10%)
Each
Chemical Identifiers
CAS12024-22-5
IUPAC Namedigallium(3+) trisulfanediide
Molecular FormulaGa2S3
InChI KeyBVSHTEBQPBBCFT-UHFFFAOYSA-N
SMILES[S--].[S--].[S--].[Ga+3].[Ga+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Gallium(III) sulfide is used in electronics and photonics. It acts as an active material in lithium secondary batteries. Further, it is used in thin-film solar cells. It is also used to prepare gallium lanthanum sulfide glass by reacting with lanthanum sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only