Lithium bromide, anhydrous, 99% min
Lithium bromide, anhydrous, 99% min
Lithium bromide, anhydrous, 99% min
Lithium bromide, anhydrous, 99% min
Thermo Scientific Chemicals

Lithium bromide, anhydrous, 99% min

CAS: 7550-35-8 | BrLi
Have Questions?
Catalog NumberQuantity
013408.0E
also known as 013408-0E
2500 g
Catalog number 013408.0E
also known as 013408-0E
Price (USD)
563.65
Special offer
Online exclusive
Ends: 28-Dec-2026
663.00
Save 99.35
Each
Quantity:
2500 g
Price (USD)
563.65
Special offer
Online exclusive
Ends: 28-Dec-2026
663.00
Save 99.35
Each

Pharmaceuticals, air-conditioning, humectant, drying agent, batteries, low temperature heat-exchange mediumLithium bromide is used for reconstitution of brines, as swelling agent for proteins and as electrolyte component in lithium battery. Used as brazing and welding flux. Lithium bromide is used as catalyst for the condensation of carbonyl compounds with active methylene compounds in the absence of solvent, operating medium for air-conditioning and industrial drying system. Also used in manufacturing pharmaceuticals and alkylation process. It also plays an important role in the synthesis of dihydropyrimidinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Pharmaceuticals, air-conditioning, humectant, drying agent, batteries, low temperature heat-exchange medium
Specifications
Boiling Point1265°C
CAS7550-35-8
Melting Point550°C
Molecular FormulaBrLi
Percent Purity≥99%
Physical FormSolid
Quantity2500 g
Unit SizeEach