Lithium carbonate, 99% - FAQs
View additional product information for Lithium carbonate, 99% - FAQs (013418.A3, 013418.A9, 013418.36)
1 product FAQs found
Is Lithium carbonate, 99% (Cat. No. 013418.36) in powder form?
Yes, Lithium carbonate, 99% (Cat. No. 013418.36) is in powder form.