Lithium molybdenum oxide, 99+%
Lithium molybdenum oxide, 99+%
Lithium molybdenum oxide, 99+%
Thermo Scientific Chemicals

Lithium molybdenum oxide, 99+%

CAS: 13568-40-6 | Li2MoO4 | 173.826 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 013427.22
also known as 013427-22
Price (USD)
64.30
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
64.30
Each
Chemical Identifiers
CAS13568-40-6
IUPAC Namedilithium(1+) dioxomolybdenumbis(olate)
Molecular FormulaLi2MoO4
InChI KeyNMHMDUCCVHOJQI-UHFFFAOYSA-N
SMILES[Li+].[Li+].[O-][Mo]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCalcium: Report
Water ContentMoisture: Report
Impurity contentAmmonia: ≤0.0005%
Impurity contentIron: ≤5 ppm
Impurity contentSulfate: ≤0.01%
View more
Lithium molybdenum oxide is mainly used as an inhibitor in some types of industrial air conditioning. It is used in comparative property studies of battery systems. It has been used as a petroleum cracking catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only