Sodium phosphate, tribasic, anhydrous, tech.
Sodium phosphate, tribasic, anhydrous, tech.
Sodium phosphate, tribasic, anhydrous, tech.
Thermo Scientific Chemicals

Sodium phosphate, tribasic, anhydrous, tech.

CAS: 7601-54-9 | Na3O4P | 163.939 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 013438.A7
also known as 013438-A7
Price (USD)
169.65
Online Exclusive
188.00
Save 18.35 (10%)
Each
Quantity:
5 kg
Request bulk or custom format
Price (USD)
169.65
Online Exclusive
188.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS7601-54-9
IUPAC Nametrisodium phosphate
Molecular FormulaNa3O4P
InChI KeyRYFMWSXOAZQYPI-UHFFFAOYSA-K
SMILES[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0% to ≤104.0%
Sodium phosphate, tribasic is used in soaps, detergents, photographic developers, water softeners and boiler scale removers. It is used as a food additive, as cleaning agents, as a protein precipitant and an active ingredient in fluxes designed to deoxygenate nonferrous metals for casting. It is used to prepare chlorinated trisodium phosphate by reacting with sodium hypochlorite and used as a disinfectant and bleach. Further, it is used in dairy substitutes and to inhibit mold. In addition, it is employed in ceramic production. It is also used for the preservation of biologically active soil samples and fractionization of nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only