Thermo Scientific Chemicals

Potassium acetate, ACS, 99% min, Thermo Scientific Chemicals

Catalog number: 013449.F7
5 x 2 kg, Each
Thermo Scientific Chemicals

Potassium acetate, ACS, 99% min, Thermo Scientific Chemicals

Catalog number: 013449.F7
5 x 2 kg, Each
Quantity
Catalog number: 013449.F7
Price (USD)
Price: 623.00
Online price: 529.65
Your price:
Quantity
-

Chemical Identifiers

CAS
127-08-2
IUPAC Name
potassium acetate
Molecular Formula
C2H3KO2
InChI Key
SCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES
[K+].CC([O-])=O
Assay (unspecified)
99% C₂H₃KO₂ min
Heavy Metals
(ICP-OES) 5ppm
Impurity content
Cl 0.003%
Impurity content
Fe 5ppm
Impurity content
Mg 0.002%

Description

Potassium acetate is used as deicer to remove snow and ice from the surface and is considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive; as a buffer in biological research; as extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins thereby removal of proteins from DNA. It plays an important role for preserving tissues, fixation and mummification. .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text