Stainless Steel rod, 6.35mm (0.25 in.) dia., Type 303, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Stainless Steel rod, 6.35mm (0.25 in.) dia., Type 303, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
013473.BV
also known as 013473-BV
30 cm
Catalog number 013473.BV
also known as 013473-BV
Price (USD)
74.70
Each
Quantity:
30 cm
Request bulk or custom format
Price (USD)
74.70
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeFe:Cr:Ni; 73:18:9 wt%
Chemical Name or MaterialStainless Steel rod, Type 303
MDL NumberMFCD00148499
Name Note6.35mm (0.25 in.) dia.
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only