Rubidium iodide, 99.8% (metals basis)
Rubidium iodide, 99.8% (metals basis)
Rubidium iodide, 99.8% (metals basis)
Thermo Scientific Chemicals

Rubidium iodide, 99.8% (metals basis)

CAS: 7790-29-6 | IRb | 212.372 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 013497.18
also known as 013497-18
Price (USD)
228.65
Online Exclusive
254.00
Save 25.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
228.65
Online Exclusive
254.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS7790-29-6
IUPAC Namerubidium(1+) iodide
Molecular FormulaIRb
InChI KeyWFUBYPSJBBQSOU-UHFFFAOYSA-M
SMILES[Rb+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Rubidium iodide is used for making metal rubidium and other rubidium salts. It is also used in the production of special glass, miniature high energy battery and crystal scintillation counter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only