Rubidium formate hydrate, 99.8% (metals basis), Thermo Scientific Chemicals
Rubidium formate hydrate, 99.8% (metals basis), Thermo Scientific Chemicals
Rubidium formate hydrate, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rubidium formate hydrate, 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
013499.09
also known as 013499-09
10 g
Catalog number 013499.09
also known as 013499-09
Price (USD)
255.65
Special offer
Online exclusive
Ends: 30-Jun-2026
301.00
Save 45.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
255.65
Special offer
Online exclusive
Ends: 30-Jun-2026
301.00
Save 45.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS3495-35-0
Chemical Name or MaterialRubidium formate hydrate
ColorWhite
Formula Weight130.49 (Anhydrous)
View more
Rubidium formate hydrate is used in anticoagulants, dental materials, indicators and reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only