Copper single crystal disc, 10mm (0.39in) dia, 1-3mm (0.04-0.1in) thick, (100) orientation
Copper single crystal disc, 10mm (0.39in) dia, 1-3mm (0.04-0.1in) thick, (100) orientation
Copper single crystal disc, 10mm (0.39in) dia, 1-3mm (0.04-0.1in) thick, (100) orientation
Thermo Scientific Chemicals

Copper single crystal disc, 10mm (0.39in) dia, 1-3mm (0.04-0.1in) thick, (100) orientation

CAS: 7440-50-8
Have Questions?
Catalog NumberQuantity
013571.KG
also known as 013571-KG
1 Ea.
Catalog number 013571.KG
also known as 013571-KG
Price (USD)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
CAS7440-50-8
Quantity1 Ea.
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.