Terbium(III) nitrate hydrate, REacton™, 99.99% (REO)
Terbium(III) nitrate hydrate, REacton™, 99.99% (REO)
Terbium(III) nitrate hydrate, REacton™, 99.99% (REO)
Terbium(III) nitrate hydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Terbium(III) nitrate hydrate, REacton™, 99.99% (REO)

CAS: 10043-27-3 | N3O9Tb | 344.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 013593.04
also known as 013593-04
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.90
Save 11.25 (15%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.90
Save 11.25 (15%)
Each
Chemical Identifiers
CAS10043-27-3
IUPAC Nameterbium(3+) trinitrate
Molecular FormulaN3O9Tb
InChI KeyYJVUGDIORBKPLC-UHFFFAOYSA-N
SMILES[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Terbium(III) nitrate hydrate is used in ceramics, glass, phosphors, lasers, and also is the important dopant for fibre amplifiers. It is also used in alloys and in the production of electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only