Sodium iodide, ultra dry, 99.999% (metals basis)
Sodium iodide, ultra dry, 99.999% (metals basis)
Sodium iodide, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals

Sodium iodide, ultra dry, 99.999% (metals basis)

CAS: 7681-82-5 | INa | 149.894 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 013604.22
also known as 013604-22
Price (USD)
684.65
Online Exclusive
760.00
Save 75.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
684.65
Online Exclusive
760.00
Save 75.35 (10%)
Each
Chemical Identifiers
CAS7681-82-5
IUPAC Namesodium iodide
Molecular FormulaINa
InChI KeyFVAUCKIRQBBSSJ-UHFFFAOYSA-M
SMILES[Na+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water< 100 ppm
Particle Size-10 Mesh
Appearance (Color)White
FormBeads
Total Metal Impurities0.001% max.
Sodium iodide commonly is used to prevent iodine deficiency. Used as analytical reagents, in the manufacture of disinfectants, antiseptics, deodorants and medicines. They are used in baking ingredient conditioner and as animal feed. It is used for conversion of alkyl chlorides into alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only