Neodymium Boron Iron, REacton™
Thermo Scientific Chemicals

Neodymium Boron Iron, REacton™

Nd:B:Fe; 15:8:77 atomic%
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
2 x 500 g
Catalog number 013642.18
also known as 013642-18
Price (USD)
80.65
Online exclusive
89.10
Save 8.45
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
80.65
Online exclusive
89.10
Save 8.45
Each
Specifications
AppearanceSilver gray broken lump
Chemical Name or MaterialNeodymium Boron Iron
ColorGray-Silver
Density7.5g/cm3
MDL NumberMFCD02091729
View more
Neodymium Boron Iron applied to manufacture head actuators for computer hard disks, magnetic resonance imaging (MRI), magnetic guitar pickups, loudspeakers and headphones, mechanical e-cigarette firing switches and magnetic bearings and couplings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only