Scandium(III) carbonate hydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Scandium(III) carbonate hydrate, REacton™, 99.99% (REO)

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 013646.03
also known as 013646-03
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Specifications
Assay Percent Notes(REO)
Chemical Name or MaterialScandium(III) carbonate hydrate
Formula Weight269.94 (Anhydrous)
MDL NumberMFCD00011501
Melting Point>500°C (decomposition)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only