Gadolinium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Gadolinium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Gadolinium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Gadolinium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Gadolinium(III) fluoride, anhydrous, REacton™, 99.9% (REO)

CAS: 13765-26-9 | F3Gd | 214.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 013655.22
also known as 013655-22
Price (USD)
387.65
Online Exclusive
430.00
Save 42.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
387.65
Online Exclusive
430.00
Save 42.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS13765-26-9
IUPAC Namegadolinium(3+) trifluoride
Molecular FormulaF3Gd
InChI KeyTYIZUJNEZNBXRS-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Gd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only