Europium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Europium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Europium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Europium(III) fluoride, anhydrous, REacton™, 99.9% (REO)

CAS: 13765-25-8 | EuF3 | 208.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 013659.04
also known as 013659-04
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS13765-25-8
IUPAC Nameeuropium(3+) trifluoride
Molecular FormulaEuF3
InChI KeyHPNURIVGONRLQI-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Eu+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Europium(III) fluoride is used as a catalyst in the preparation of novel mixed-metal fluorides. It finds application in studies related to fluoride glasses. It is also used as a laboratory reagent. Further, it is used in optical glasses and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only