Cobalt(II) chloride, ultra dry, 99.998% (metals basis)
Cobalt(II) chloride, ultra dry, 99.998% (metals basis)
Cobalt(II) chloride, ultra dry, 99.998% (metals basis)
Cobalt(II) chloride, ultra dry, 99.998% (metals basis)
Thermo Scientific Chemicals

Cobalt(II) chloride, ultra dry, 99.998% (metals basis)

Hygroscopic; 3.367. Decomposes at 400°C on long heating in air | CAS: 7646-79-9 | Cl2Co | 129.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 013665.03
also known as 013665-03
Price (USD)
58.65
Online Exclusive
64.70
Save 6.05 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
58.65
Online Exclusive
64.70
Save 6.05 (9%)
Each
Add to cart
Chemical Identifiers
CAS7646-79-9
IUPAC Nameλ²-cobalt(2+) dichloride
Molecular FormulaCl2Co
InChI KeyGVPFVAHMJGGAJG-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-10 Mesh
FormBeads
Appearance (Color)Blue
Total Metal Impurities0.002% max.
Residual water< 100 ppm
Cobalt(II) chloride is used in humidity indicator in weather instruments. In the anhydrous form, it finds use in electroplating of cobalt, in organic chemistry and is a precursor to cobaltocene, (bis(cyclopentadienyl)cobalt(II), which is a good reducing agent. It also serves as a Lewis acid. Cobalt chloride is an indicator for water in desiccants, owing to the reversible hydration/dehydration coupled with distinct color change. Cobalt chloride is useful for producing invisible ink as it turns blue when heated and becomes invisible once it gets cooled. Cobalt(II) chloride catalyzes cross coupling of aryl halides or vinyl halides with aryl grignard reagents in excellent yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only